Cefotaxime
Catalog # PROOTRCC242958-250
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:455.46 g/mol
Melting Pt:>158 °C
:GHS08
CAS Number:63527-52-6
Formula:C₁₆H₁₇N₅O₇S₂
MDL Number:MFCD00864971