Chrysoeriol
Catalog # PROOTRCQ509540-2.5
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:2,5 mg
- CAS No.:18609-17-1
- Description:Quercetin-3-O-sophoroside
- Shipping conditions:RT
- Formula:C₂₇H₃₀O₁₇
- Storage temperature:−20 °C
- Molecular weight (Mw):626.517
- Volume:2,5 mg