ContactUsLinkComponent
Cell Explorer™ Live Cell Tracking Kit
Cell Explorer™ Live Cell Tracking Kit
Catalog # AATB22620
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cell Explorer™ Live Cell Tracking Kit
Catalog # AATB22620
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    Kit is designed to uniformly label live cells in blue fluorescence for the studies that require the fluorescent tag molecules retained inside cells for relatively longer time
Specifications
About this item
The kit uses a weakly fluorescent dye that carries a cell-retaining moiety.
  • Long term tracking
  • Suitable for variety of studies
  • Optimized cell labeling protocol
The labeling process is robust, requiring minimal hands-on time. It can be readily adapted for a wide variety of fluorescence platforms such as microplate assays, immunocytochemistry and flow cytometry.
Packaging: Plastic vial.
Caution: For research use only.