Order Entry
Germany
ContactUsLinkComponent
ReadiLink™ Rapid XFD488 Antibody Labelling Kit 'XFD488 Same Structure to Alexa Fluor™ 488' (2×50 µg Labellings)
ReadiLink™ Rapid XFD488 Antibody Labelling Kit 'XFD488 Same Structure to Alexa Fluor™ 488' (2×50 µg Labellings)
Catalog # AATB1275
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
ReadiLink™ Rapid XFD488 Antibody Labelling Kit 'XFD488 Same Structure to Alexa Fluor™ 488' (2×50 µg Labellings)
Catalog # AATB1275
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Description:
    ReadiLink™ rapid XFD488 antibody labeling kit 'XFD488 same structure to Alexa Fluor™ 488'

Specifications

About this item

XFD488 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 488 (Alexa Fluor® is the trademark of ThermoFisher).

  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labelling protocol
  • No column purification required
Caution: For research use only.