ReadiLink™ xtra Rapid iFluor® 647 Antibody Labelling Kit *BSA-Compatible* (2 Labellings)
Artikelnr. AATB1963
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:2 Tests
- Description:ReadiLink™ xtra rapid iFluor™ 647 antibody labeling kit 'BSA-compatible'
Specifications
About this item
ReadiLink™ xtra rapid antibody labelling kits require essentially only 2 simple mixing steps without a column purification needed.
- Contains all essential components
- Minimal hands on time
- Ready to use
- Robust and simple antibody labelling protocol
- No column purification required
Caution: For research use only.