ContactUsLinkComponent
Pluronic® F-127 Cell Culture Tested
Pluronic® F-127 Cell Culture Tested
Catalog # AATB20050
Supplier:  AAT BIOQUEST
undefined
Pluronic® F-127 Cell Culture Tested
Catalog # AATB20050
Supplier:  AAT BIOQUEST
Specifications
  • Pk:
    10 g
Specifications
About this item
Pluronic® F-127 is a nonionic surfactant that is 100% active and relatively non-toxic to cells, and frequently used with dye AM esters to improve their water solubility.
  • Non-toxic to cells
It is soluble at low temperature, but gels at room temperature.