Helixyte™ Green *10000X Aqueous PCR Solution*
Catalog # AATB17592
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:1 mL
- Description:Helixyte™ Green
- Purity:95%
Specifications
About this item
Helixyte™ Green is manufactured by AAT Bioquest, and it has the same chemical structure of SYBR® Green (SYBR® is the trademark of ThermoFisher).
- Same Spectral Properties To SYBR® Green
- High Sensitivity
- Useful For Nucleic Acid Labeling In Gels
Caution: For research use only.