Cell Meter™ Fluorimetric Cellular Lipid Peroxidation Assay Kit
Catalog # AATB22906
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:200 Tests
- Description:Cell Meter™ Fluorimetric cellular lipid peroxidation assay kit
- Tests per kit:200 Tests
Specifications
About this item
Lipid peroxidation is the oxidative degradation of cellular lipid by reactive oxygen species (ROS).
- Contains all necessary components
- Multiple research applications
- Ratiometric detection
- Suitable for all major fluorescence platforms
Caution: For research use only.