Cell Meter™ Autophagy Assay Kit *Blue Fluorescence*
Catalog # AATB23000
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:200 Tests
- Description:Cell Meter™ Autophagy assay kit *Blue fluorescence*
- Tests per kit:200 Tests
Specifications
About this item
Autophagy is an evolutionarily conserved degradation process that targets long-lived proteins, organelles, and other cytoplasmic components for degradation via the lysosomal pathway.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Suitable for either detached or attached cells
Caution: For research use only.