Spexyte™ Micro pH Probe *Optimized for Nanodrop*
Catalog # AATB21230
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:500 Tests
- Description:Spexyte™ Micro pH probe *Optimized for Nanodrop*
- Tests per kit:500 Tests
Specifications
About this item
Biological samples are usually in a very narrow pH range from 4.5 to 8.5, and many of them need to be provided in micro-volume (microliter).
- Suitable for micro-volume samples
- Sensitive and quick results
- Fits four-parameter logistic model
Caution: For research use only.