Cell Meter™ Mitochondrion Membrane Potential Assay Kit *Orange Fluorescence Optimized for Microplate Reader*
Cell Meter™ mitochondrion membrane potential assay kit *orange fluorescence optimized for microplate reader*, 500 Tests, 500 Tests
Catalog # AATB22805
Supplier: AAT Bioquest
Cell Meter™ Mitochondrion Membrane Potential Assay Kit *Orange Fluorescence Optimized for Microplate Reader*
Catalog # AATB22805
Supplier: AAT Bioquest
CAS Number:
Jump to:
Product Details & Documents
Our Cell Meter™ assay kits are a set of tools for monitoring cellular functions.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Optimized for microplate readers but suitable for all major fluorescence platforms
Caution: For research use only.
Specifications
Specifications
- Pk:500 Tests
- Description:Cell Meter™ mitochondrion membrane potential assay kit *orange fluorescence optimized for microplate reader*
- Tests per kit:500 Tests



