Annexin V-iFluor® 555 Conjugate
Catalog # AATB20072
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:100 Tests
- Description:Annexin V-iFluor™ 555 conjugate
- Tests per kit:100 Tests
Specifications
About this item
Annexins are a family of proteins that bind to phospholipid membranes in the presence of calcium.
- High stability
- In-house quality control
- Multiple research applications
Caution: For research use only.