Order Entry
Germany
Orders LinkContactUsLinkComponent
LysoBrite™ Deep Red

LysoBrite™ Deep Red

LysoBrite™ Deep Red, 500 Tests, 500 Tests
Catalog # AATB22646
Supplier:   AAT Bioquest
LysoBrite™ Deep Red
Catalog # AATB22646
Supplier:  AAT Bioquest
CAS Number:  
LysoBrite™ Deep Red
LysoBrite™ Deep Red

LysoBrite™ Deep Red, 500 Tests, 500 Tests

CAS Number:  
Price
235,00 €
Log in for availability
1 * 500 Tests
Product Details & Documents

Lysosomes are cellular organelles which contain acid hydrolase enzymes to break up waste materials and cellular debris.

  • Lysosome-Specific Dye
  • Long-Term Use
  • Low Cytotoxicity

Caution: For research use only.

Specifications

Specifications

  • Pk:
    500 Tests
  • Description:
    LysoBrite™ Deep Red
  • Tests per kit:
    500 Tests
  • Cat. No.:
    AATB22646
  • Purity:
    95%

Specifications