LysoBrite™ Blue
Catalog # AATB22642
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:500 Tests
- Description:LysoBrite™ Blue
- Purity:90%
- Tests per kit:500 Tests
Specifications
About this item
Lysosomes are cellular organelles which contain acid hydrolase enzymes to break up waste materials and cellular debris.
- Lysosome-Specific Dye
- Long-Term Use
- Low Cytotoxicity
Caution: For research use only.