Orders LinkContactUsLinkComponent
Cell Meter™ Fixable Viability Dye, RX660
Cell Meter™ Fixable Viability Dye, RX660
Cell Meter™ fixable viability dye, 200 Tests, 200 Tests
Artikelnummer AATB22530
Hersteller:   AAT Bioquest
Cell Meter™ Fixable Viability Dye, RX660
Artikelnummer AATB22530
Hersteller:  AAT Bioquest
CAS-Nummer:  
Cell Meter™ Fixable Viability Dye, RX660
Cell Meter™ Fixable Viability Dye, RX660
Cell Meter™ fixable viability dye, 200 Tests, 200 Tests
CAS-Nummer:  
Preis
299,00 €
Log in for availability
1 * 200 Tests
Product Details & Documents
Discrimination and exclusion of dead cells from live cells allows cleaner separation and identification of cell populations.
  • Cell-Impermeable
  • Amine- And Thiol-Reactive
  • High Stability
  • Live/Dead Cell Labeling Dye
  • Fixable
Caution: For research use only.
Specifications
Specifications
  • Pk:
    200 Tests
  • Description:
    Cell Meter™ fixable viability dye
  • Tests per kit:
    200 Tests