Order Entry
Germany
Orders LinkContactUsLinkComponent
Nuclear Red™ LCS2

Nuclear Red™ LCS2

Nuclear Red™ LCS2, 0,5 mL
Catalog # AATB17545
Supplier:   AAT Bioquest
Nuclear Red™ LCS2
Catalog # AATB17545
Supplier:  AAT Bioquest
CAS Number:  
Nuclear Red™ LCS2
Nuclear Red™ LCS2

Nuclear Red™ LCS2, 0,5 mL

CAS Number:  
Price
156,00 €
Log in for availability
1 * 0,5 mL
Product Details & Documents

Our Nuclear Red™ LCS2 is a fluorogenic, DNA-selective and cell-permeant dye for analyzing DNA content in living cells.

  • Cell-Permeable
  • Useful For Multicolor Imaging
  • Bright Fluorescence

5 mM DMSO Solution

Caution: For research use only.

Specifications

Specifications

  • Pk:
    0,5 mL
  • Description:
    Nuclear Red™ LCS2
  • Cat. No.:
    AATB17545
  • Purity:
    90%

Specifications