Nuclear Orange™ LCS1
Catalog # AATB17541
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:0,5 mL
- Description:Nuclear Orange™ LCS1
- Purity:95%
Specifications
About this item
The Nuclear Orange™ LCS1 is a fluorogenic, DNA-selective and cell-permeant dye for analyzing DNA content in living cells.
- Cell-Permeable
- Useful For Multicolor Imaging
- Bright Fluorescence
5 mM DMSO Solution
Caution: For research use only.