Order Entry
Germany
ContactUsLinkComponent
 
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Alexa Fluor® 594)
  AATB16388
undefined
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Alexa Fluor® 594)
  AATB16388

 

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Clone:
    IgG
  • Conjugation:
    Alexa Fluor® 594
  • Host:
    Goat
  • Isotype:
    IgG (H, L)
  • Reactivity:
    Mouse
  • Cross adsorption:
    Yes
  • Size:
    1 mg
  • Pack type:
    Vial
  • Pk:
    1 mg

 

 

XFD594 has the same chemical structure of Alexa Fluor® 594 (Alexa Fluor® is the trademark of ThermoFisher).

  • Affinity-purified
  • Bright signal
  • Cross-adsorbed

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: Alexa Fluor® 594
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse