Polyethylene Heavy-Duty Graduated Buckets
Supplier: LAMAPLAST
Certificates
About this item
Made with low density polyethylene (LDPE), in white or black colour options.
- Conical shape, litre graduations
- With ergonomic handle
- Food contact material approved
Buckets feature a flat bottom design to ensure total emptying and allow for easy cleaning.
Certifications: EU 10/2011.