0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Pyranine
Pyranine ≥95%
Pyranine ≥95%
AATB85
:
AAT BIOQUEST
:
Pyranine ≥95%
AATB85
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
1 g
Seq:
3047
:
:
5,II
:
524.371 g/mol
:
GHS07
:
6358-69-6
:
C₁₆H₇Na₃O₁₀S₃
:
MFCD00037575