Pyronin G ≥95%
Plastic bottle, 100 mg
Catalog # AATB61
Supplier: AAT Bioquest
Jump to:
Product Details & Documents
Pyronin Y is widely used in combination with methyl green to selectively and differentially stain nucleic acids following polyacrylamide gel electrophoresis.
Synonyms: Pyronin Y, Pyronine Y, 3,6-Bis(dimethylamino)xanthylium chloride
- CAS Number:92-32-0
- Formula:C₁₇H₁₉ClN₂O
- MDL Number:MFCD00011725
- Molecular Weight:302.8 g/mol
- ADR:5,II
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:100 mg



