2-Aminoacridone ≥95%
Plastic bottle, 25 mg
Catalog # AATB604
Supplier: AAT Bioquest
Jump to:
Product Details & Documents
This reagent is used to modify carbonyl-containing biomolecules, in particular, for labeling carbohydrates and glycoproteins.
Synonyms: 2-Aminoacridin-9(10H)-one
- CAS Number:27918-14-5
- Formula:C₁₃H₁₀N₂O
- MDL Number:MFCD00037397
- Molecular Weight:246.69 g/mol
- ADR:5,II
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:25 mg



