Order Entry
Germany
ContactUsLinkComponent
Texas Red-X, succinimidyl ester mixture of isomers ≥90%
Irritant
Texas Red-X, succinimidyl ester mixture of isomers ≥90%
Catalog # AATB475
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
Texas Red-X, succinimidyl ester mixture of isomers ≥90%
Catalog # AATB475
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :816.94 g/mol
ADR:5,II
 :GHS07
CAS Number:216972-99-5
Formula:C₄₁H₄₄N₄O₁₀S₂