Order Entry
Germany
ContactUsLinkComponent
Cyanine 5 monosuccinimidyl ester potassium salt ≥90% [same as GE Cy5® NHS ester]
Cyanine 5 monosuccinimidyl ester potassium salt ≥90% [same as GE Cy5® NHS ester]
Catalog # AATB280
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cyanine 5 monosuccinimidyl ester potassium salt ≥90% [same as GE Cy5® NHS ester]
Catalog # AATB280
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :791.97 g/mol
CAS Number:146368-14-1