0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
N-Sp-DMAE NHS ester
N-Sp-DMAE NHS ester for biomolecular labeling
N-Sp-DMAE NHS ester for biomolecular labeling
:
AAT BIOQUEST
New Product
N-Sp-DMAE NHS ester for biomolecular labeling
:
AAT BIOQUEST
New Product
:
194357-64-7
:
:
590.6 g/mol
:
5,II
:
GHS07