Order Entry
Germany
Orders LinkContactUsLinkComponent
6-HEX azide ≥95%

6-HEX azide ≥95%

5 mg
Catalog # AATB240
Supplier:   AAT Bioquest
6-HEX azide ≥95%
Catalog # AATB240
Supplier:  AAT Bioquest
CAS Number:  
6-HEX azide ≥95%
6-HEX azide ≥95%

5 mg

CAS Number:  
Price
235,00 €
Log in for availability
1 * 5 mg
Product Details & Documents

6-HEX is widely used in nucleic acid sequencing and related research.

  • Multiple Bioconjugation Applications
  • Excellent Quality

Caution: For research use only.

  • Molecular Weight:651.07 g/mol
Specifications

Specifications

  • Pk:
    5 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.