Order Entry
Germany
Orders LinkContactUsLinkComponent
RH421 ≥90%

RH421 ≥90%

Plastic bottle, 5 mg
Catalog # AATB21482
Irritant
Supplier:   AAT Bioquest
RH421 ≥90%
Catalog # AATB21482
Supplier:  AAT Bioquest
CAS Number:  107610-19-5
RH421 ≥90%
RH421 ≥90%

Plastic bottle, 5 mg

CAS Number:  107610-19-5
Price
156,00 €
Log in for availability
1 * 5 mg
Product Details & Documents

RH 421, also called N-(4-Sulfobutyl)-4-(4-(4- (dipentylamino)phenyl)butadienyl)pyridinium, is a neuron-tracing dye.

Synonyms:  4-{4-[4-(Dipentylamino)phenyl]-1,3-butadienyl}-1-(4-sulphobutyl)pyridinium hydroxide, N-(4-sulphobutyl)-4-(4-(4-(dipentylamino)phenyl)butadienyl) pyridinium, inner salt
  • CAS Number:107610-19-5
  • Formula:C₂₉H₄₂N₂O₃S
  • Molecular Weight:498.72 g/mol
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.