Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Rhod-4™ potassium salt
Rhod-4™ potassium salt
Rhod-4™ potassium salt
Rhod-4™ potassium salt
Supplier:
AAT BIOQUEST
Certificates
Expand image
Rhod-4™ potassium salt
Supplier:
AAT BIOQUEST
About this item
:
:
880.07 g/mol
ADR:
5,II
:
GHS07