Order Entry
Germany
Orders LinkContactUsLinkComponent
 

FLUO 3/AM

Plastic bottle, 50 mg
Catalog # AATB21012
Irritant
Supplier:   AAT Bioquest
FLUO 3/AM
Catalog # AATB21012
Supplier:  AAT Bioquest
CAS Number:  121714-22-5
FLUO 3/AM
FLUO 3/AM

Plastic bottle, 50 mg

CAS Number:  121714-22-5
Price
6 240,00 €
Log in for availability
1 * 50 mg
Product Details & Documents

Calcium measurement is critical for numerous biological investigations.

Synonyms:  4-(6-Acetoxymethoxy-2,7-dichloro-3-oxo-9-xanthenyl)-4′-methyl-2,2′(ethylenedioxy)dianiline-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester, 1-[Amino-5-(2,7-dichloro-6-acetomethoxy-3-oxo-3H-xanthen-9-yl)phenoxy]-2-(2'-amino-5'-methylphenoxy)ethane-N,N,N',N'-tetraacetic acid pentaacetoxy-methyl ester
  • CAS Number:121714-22-5
  • Formula:C₅₁H₅₀Cl₂N₂O₂₃
  • Molecular Weight:1129.85 g/mol
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 mg

Specifications