0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
DNP-PEG4-NHS ester
DNP-PEG4-NHS ester FRET quencher precursor
DNP-PEG4-NHS ester FRET quencher precursor
AATB2023
:
AAT BIOQUEST
:
DNP-PEG4-NHS ester FRET quencher precursor
AATB2023
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
5 mg
Seq:
2210
:
:
528.467 g/mol
:
5,II
:
GHS07
:
858126-78-0
:
C₂₁H₂₈N₄O₁₂
:
MFCD13184987