0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
XFD546 NHS Ester
XFD546 NHS Ester fluorescent probe
Irritant
XFD546 NHS Ester fluorescent probe
Catalog # AATB1820
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
Irritant
XFD546 NHS Ester fluorescent probe
Catalog # AATB1820
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
1159.6 g/mol
ADR:
5,II
:
GHS07
Product Details