Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
bisBenzimide H-33342 (Hoechst 33342)
bisBenzimide H-33342 (Hoechst 33342) 20 mM in water ≥95%
bisBenzimide H-33342 (Hoechst 33342) 20 mM in water ≥95%
bisBenzimide H-33342 (Hoechst 33342) 20 mM in water ≥95%
Catalog # AATB17535
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
bisBenzimide H-33342 (Hoechst 33342) 20 mM in water ≥95%
Catalog # AATB17535
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pk:
5 mL
Seq:
2210
Specifications
About this item
Product Details
:
:
452.562 g/mol
CAS Number:
23491-52-3
Formula:
C₂₇H₂₈N₆O
Product Details