Orders LinkContactUsLinkComponent
DAPI Dilactate ≥95%
DAPI Dilactate ≥95%
25 mg
Catalog # AATB17509
Supplier:   AAT Bioquest
DAPI Dilactate ≥95%
Catalog # AATB17509
Supplier:  AAT Bioquest
CAS Number:  28718-91-4
DAPI Dilactate ≥95%
DAPI Dilactate ≥95%
25 mg
CAS Number:  28718-91-4
Price
156,00 €
Log in for availability
1 * 25 mg
Product Details & Documents
DAPI is a fluorescent stain that binds strongly to DNA.
  • Binding-Determined Bicolor Fluorescence
  • Cell-Permeable
  • Useful For Multicolor Imaging
Caution: For research use only.
Synonyms:  2-(4-Carbamimidoylphenyl)indole-6-carboximidamide dilactate, 2-(4-Amidinophenyl)indole-6-carboxamidine dilactate
  • CAS Number:28718-91-4
Specifications
Specifications
  • Pk:
    25 mg
Specifications