Order Entry
Germany
Orders LinkContactUsLinkComponent
Biotin-11-dATP

Biotin-11-dATP

Plastic bottle, 25 Nmo
Catalog # AATB17014
Irritant
Supplier:   AAT Bioquest
Biotin-11-dATP
Catalog # AATB17014
Supplier:  AAT Bioquest
CAS Number:  
Biotin-11-dATP
Biotin-11-dATP

Plastic bottle, 25 Nmo

CAS Number:  
Price
147,00 €
Log in for availability
1 * 25 Nmo
Product Details & Documents

The biotin-modified deoxyadenosine 5'-triphosphates are widely used for a variety of non-radioactive DNA labeling reactions including nick translation, random prime labeling, cDNA labeling and 3’-end labeling.

  • Useful for DNA probe development
  • Well-cited
  • Multiple applications

Caution: For research use only.

  • Molecular Weight:882.71 g/mol
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 Nmo

Specifications