Fluorescein di-b-D-galactopyranoside (FDG) ≥95%
Catalog # AATB14001
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:656.593 g/mol
ADR:5,II
:GHS07
CAS Number:17817-20-8
Formula:C₃₂H₃₂O₁₅