Order Entry
Germany
ContactUsLinkComponent
Cyanine 3.5 amine ≥95% [equivalent to Cy3.5® amine]
Cyanine 3.5 amine ≥95% [equivalent to Cy3.5® amine]
Catalog # AATB139
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cyanine 3.5 amine ≥95% [equivalent to Cy3.5® amine]
Catalog # AATB139
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :1161.15 g/mol