Order Entry
Germany
ContactUsLinkComponent
iFluor® 568 Tris NTA-Ni complex, HIS Lite™
Irritant
iFluor® 568 Tris NTA-Ni complex, HIS Lite™
Catalog # AATB12617
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
iFluor® 568 Tris NTA-Ni complex, HIS Lite™
Catalog # AATB12617
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 µG
  • Seq:
    1286

Specifications

About this item

 :
ADR:5,II
 :GHS07