Order Entry
Germany
ContactUsLinkComponent
NTA succinimidyl ester
NTA succinimidyl ester
Catalog # AATB12608
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
NTA succinimidyl ester
Catalog # AATB12608
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :515.52 g/mol
ADR:5,II
 :GHS07