D(-)-Luciferin ≥95%
Supplier: AAT Bioquest
Jump to:
About this item
Luciferin is the most popular and versatile bioluminescent substrate.
Synonyms: D-Luciferin, (S)-2-(6-Hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid
- CAS Number:2591-17-5
- Formula:C₁₁H₈N₂O₃S₂
- MDL Number:MFCD00042929
- Molecular Weight:280.3228 g/mol
- ADR:5,II
3 Options Available Below
Product Details & Documents
Luciferin is the most popular and versatile bioluminescent substrate.
Synonyms: D-Luciferin, (S)-2-(6-Hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid
- CAS Number:2591-17-5
- Formula:C₁₁H₈N₂O₃S₂
- MDL Number:MFCD00042929
- Molecular Weight:280.3228 g/mol
- ADR:5,II
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



