Order Entry
Germany
Orders LinkContactUsLinkComponent
SunRed™ Acetate

SunRed™ Acetate

Plastic bottle, 25 mg
Catalog # AATB11405
Irritant
Supplier:   AAT Bioquest
SunRed™ Acetate
Catalog # AATB11405
Supplier:  AAT Bioquest
CAS Number:  
SunRed™ Acetate
SunRed™ Acetate

Plastic bottle, 25 mg

CAS Number:  
Price
156,00 €
Log in for availability
1 * 25 mg
Product Details & Documents

Esterase-catalyzed hydrolysis of Sun Red acetate (SRA) yields the Sun Red fluorophore that can be excited with the 633 nm laser with emission of ~660 nm.

  • Optimised for Cy5 filter set
  • Extremely sensitive NIR esterase substrate
  • Excellent cell viability stain
  • Useful in combination with green vital stains
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.