Order Entry
Germany
ContactUsLinkComponent
iFluor® 700, Hydrazide
iFluor® 700, Hydrazide
Catalog # AATB1087
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
iFluor® 700, Hydrazide
Catalog # AATB1087
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
ADR:5,II
 :GHS07