Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
iFluor®
iFluor® 460, Maleimide fluorescent probe
iFluor® 460, Maleimide fluorescent probe
iFluor® 460, Maleimide fluorescent probe
AATB1058
:
AAT BIOQUEST
:
iFluor® 460, Maleimide fluorescent probe
AATB1058
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
:
:
5,II
:
GHS07