Order Entry
Germany
ContactUsLinkComponent
2,3,4,6-Tetrachloroanisole, white
2,3,4,6-Tetrachloroanisole, white
Catalog # PROOTRCT223190-250
CAS Number:  
2,3,4,6-Tetrachloroanisole, white
Catalog # PROOTRCT223190-250
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :245.92 g/mol
Melting Pt:61...63 °C
 :GHS07
CAS Number:938-22-7
Formula:C₇H₄Cl₄O
MDL Number:MFCD00129714