Galileo thermometers
Supplier: MOLLER THERM
Certificates
About this item
Room thermometer with five (340 mm) or seven (420 mm) balls.
- Fluid's density changes with temperature
- Engraved weights hang in the partially fluid-filled balls
- Reading at the bottom of the balls that float up into the cylinder
Colour in the ordering table indicates the colour of the balls.