Filter holder with funnel, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
These graduated polysulfone filter holders can be used with any filtering flask or manifold that accepts a suitable rubber stopper with hole.
- Environmentally friendly alternative to disposable filters
Filtration area, with analytical support plate: 11,25 cm².
Filtration area, with sterilisation support plate: 13,30 cm².
Delivery: Comes with one easy-to-use, no twist vacuum gasket which can replace a No. 7 or 8 rubber stopper with 15 mm hole.
Specifications
- Catalog No:
- NALGDS0310-4050
- 513-3101
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement