Anti-Chloride Intracellular Channel 4 (CLIC4) Rabbit Polyclonal Antibody
Catalog # AVIVOAAN02134-50UL
Supplier: Aviva Systems Biology
Specifications
- Antigen name:Chloride Intracellular Channel 4
- Antigen symbol:CLIC4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Format:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Form:Liquid
- Gene ID:25932
- Antigen synonyms:H1|huH1|p64H1|CLIC4L|MTCLIC
- Storage temperature:Store at –20 ℃. Avoid repeated freeze/thaw cycles.
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1 to 253 of human CLIC4 (NP_039234.1)
- Purification:Affinity purified against immunogen
- Pk:50 µl
Specifications
About this item
Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume. Chloride intracellular channel 4 (CLIC4) protein, encoded by the CLIC4 gene, is a member of the p64 family; the gene is expressed in many tissues and exhibits a intracellular vesicular pattern in Panc-1 cells (pancreatic cancer cells).
Type:
Antigen: Chloride Intracellular Channel 4 (CLIC4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat