Canola oil
Catalog # SPCMC1523-500GM
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pack type:Glass bottle, amber
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Appearance @ 25 °C:Clear, Bright, Y
- Iodine Value:94 - 126
- Saponification Value:186 - 198
- Acid Value:max. 0.5
- Peroxide Value:max. 10
Specification Test Results
About this item
:
Density:1.00
Storage Temperature:Ambient
CAS Number:120962-03-0
MDL Number:MFCD00217791