
Nicardipine hydrochloride, yellow, solid
Catalog # PROOTRCN394500-50
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
Specification Test Results
- Crystalline powder:Appearance
- Solubility:DMSO (Slightly), Methanol (Slightly)
Specification Test Results
About this item
:
:515.99 g/mol
Melting Pt:176...178 °C
UN:2811
ADR:6.1,III
:GHS06
CAS Number:54527-84-3
Formula:C₂₆H₂₉N₃O₆
MDL Number:MFCD00057327