Aluminium Support Cups for XRF
Supplier: SPECAC
Certificates
About this item
These collapsible cups are designed to support the powder while it is under load within the die.
The aluminium deforms with the sample and results in a strong, supportive backing for the finished pellet. These cups are available in different sizes and starting shapes.