PIG® Drive-Over DrainBlocker® Drain Covers
Supplier: NEW PIG
Certificates
About this item
PIG® DrainBlocker® Covers deploy quickly and create a tight, temporary seal over drains to keep out unwanted liquids. Ideal for heavy outdoor vehicle traffic.
- Trust this drive-over drain cover - the proven-toughest on the market - to resist everything from punctures, stretching and tearing to prolonged exposure to weather, UV/IR light and oils like diesel fuel and gasoline, plus many chemicals
- Maintain a professional look for longer since this drive-over drain cover is designed to wipe clean and get back on the job
- Use it how you need it, either leaving it down as a preventive measure in leak- and spill-prone areas or storing it for emergency spill response
- Respond faster to spills by conveniently carrying the cover in the included nylon storage bag
Confidently block drains from spills and water with this outdoor drain cover's super-sealing bottom layer that conforms perfectly for a tight seal while the HogHyde® rubber top layer withstands years and years of repeated heavy vehicle traffic.
Specifications
- Catalog No:
- NEWPPLR901
- NEWPPLR906
- Storage temperature:
- Store from 0...48,8 °C
- Store from 0...48,8 °C